Assay Details
| Gene Symbol: LOC101116577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972088.1 | XP_027827889.1 | 4 - 5 | 2377 | 87 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101116577| Gene Symbol: LOC101116577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972088.1 | XP_027827889.1 | 4 - 5 | 2377 | 87 | |