Assay Details
| Gene Symbol: TASOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015099687.2 | XP_014955173.2 | 13 - 14 | 5412 | 109 | |
| XM_015099688.2 | XP_014955174.2 | 13 - 14 | 5393 | 109 | ||
| XM_015099691.2 | XP_014955177.2 | 13 - 14 | 5411 | 109 | ||
| XM_027976780.1 | XP_027832581.1 | 13 - 14 | 5412 | 109 | ||
| XM_027976781.1 | XP_027832582.1 | 14 - 15 | 1842 | 109 | ||
| XM_027976782.1 | XP_027832583.1 | 7 - 8 | 535 | 109 | ||
| XM_027976783.1 | XP_027832584.1 | 13 - 14 | 5393 | 109 | ||
More Information
Set Membership: |
|

TASOR2
Genomic Map