Assay Details
| Gene Symbol: TASOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015099687.2 | XP_014955173.2 | 15 - 16 | 6210 | 98 | |
| XM_015099688.2 | XP_014955174.2 | 15 - 16 | 6191 | 98 | ||
| XM_027976780.1 | XP_027832581.1 | 15 - 16 | 6210 | 98 | ||
| XM_027976781.1 | XP_027832582.1 | 16 - 17 | 2640 | 98 | ||
| XM_027976782.1 | XP_027832583.1 | 9 - 10 | 1333 | 98 | ||

TASOR2
Genomic Map