Assay Details
| Gene Symbol: TASOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015099687.2 | XP_014955173.2 | 5 - 6 | 4645 | 94 | |
| XM_015099688.2 | XP_014955174.2 | 5 - 6 | 4644 | 94 | ||
| XM_015099691.2 | XP_014955177.2 | 5 - 6 | 4644 | 94 | ||
| XM_027976780.1 | XP_027832581.1 | 5 - 6 | 4645 | 94 | ||
| XM_027976781.1 | XP_027832582.1 | 6 - 7 | 1075 | 94 | ||
| XM_027976783.1 | XP_027832584.1 | 5 - 6 | 4644 | 94 | ||

TASOR2
Genomic Map