Assay Details
| Gene Symbol: LOC101109993 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977924.1 | XP_027833725.1 | 10 - 11 | 3330 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101109993| Gene Symbol: LOC101109993 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977924.1 | XP_027833725.1 | 10 - 11 | 3330 | 93 | |
Set Membership: |
|