Assay Details
| Gene Symbol: SSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004611.3 | XP_004004660.1 | 2 - 3 | 182 | 67 | |
| XM_012134294.2 | XP_011989684.1 | 3 - 4 | 420 | 67 | ||
More Information
Set Membership: |
|

SSB
Genomic Map