Assay Details
| Gene Symbol: PRLH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009434.1 | NP_001009434.1 | 2 - 3 | 189 | 57 | |
| GenBank mRNA | AF450453.1 | - | 2 - 3 | 189 | 57 | |
More Information
Set Membership: |
|

PRLH
Genomic Map