Assay Details
| Gene Symbol: ABCF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012177365.3 | XP_012032755.1 | 8 - 9 | 1504 | 61 | |
| XM_015095448.2 | XP_014950934.1 | 8 - 9 | 1157 | 61 | ||
More Information
Set Membership: |
|

ABCF2
Genomic Map