Assay Details
| Gene Symbol: CCNH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004009073.4 | XP_004009122.1 | 1 - 2 | 262 | 69 | |
| XM_012178935.3 | XP_012034325.1 | 1 - 2 | 263 | 69 | ||
| XM_012178936.3 | XP_012034326.1 | 1 - 2 | 263 | 69 | ||
More Information
Set Membership: |
|

CCNH
Genomic Map