Assay Details
| Gene Symbol: BBOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016354.4 | XP_004016403.1 | 2 - 3 | 941 | 75 | |
| XM_027979575.1 | XP_027835376.1 | 3 - 4 | 772 | 75 | ||
| XM_027979576.1 | XP_027835377.1 | 2 - 3 | 941 | 75 | ||
More Information
Set Membership: |
|

BBOX1
Genomic Map