Assay Details
| Gene Symbol: LGI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020043.4 | XP_004020092.1 | 1 - 2 | 534 | 75 | |
| XM_012102587.3 | XP_011957977.1 | 1 - 2 | 539 | 75 | ||
More Information
Set Membership: |
|

LGI1
Genomic Map