Assay Details
| Gene Symbol: NUAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015099021.2 | XP_014954507.2 | 3 - 4 | 603 | 54 | |
| XM_027976211.1 | XP_027832012.1 | 3 - 4 | 4247 | 54 | ||
More Information
Set Membership: |
|

NUAK2
Genomic Map