Assay Details
| Gene Symbol: DPEP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978104.1 | XP_027833905.1 | 8 - 9 | 1113 | 72 | |
| XM_027978105.1 | XP_027833906.1 | 8 - 9 | 1119 | 72 | ||
More Information
Set Membership: |
|

DPEP2
Genomic Map