Assay Details
| Gene Symbol: PKMYT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961284.1 | XP_027817085.1 | 2 - 3 | 1228 | 69 | |
| XM_027961285.1 | XP_027817086.1 | 3 - 4 | 1148 | 69 | ||
| XM_027961286.1 | XP_027817087.1 | 3 - 4 | 1244 | 69 | ||
More Information
Set Membership: |
|

PKMYT1
Genomic Map