Assay Details
| Gene Symbol: SGTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012178107.3 | XP_012033497.1 | 9 - 10 | 852 | 57 | |
| XM_012178110.3 | XP_012033500.1 | 9 - 10 | 852 | 57 | ||
| XM_012178111.3 | XP_012033501.1 | 8 - 9 | 780 | 57 | ||
| XM_027969915.1 | XP_027825716.1 | 8 - 9 | 780 | 57 | ||
More Information
Set Membership: |
|

SGTA
Genomic Map