Assay Details
| Gene Symbol: VPS4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978125.1 | XP_027833926.1 | 10 - 11 | 1426 | 61 | |
| XM_027978126.1 | XP_027833927.1 | 10 - 11 | 1326 | 61 | ||
More Information
Set Membership: |
|

VPS4A
Genomic Map