Assay Details
| Gene Symbol: YY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957397.1 | XP_027813198.1 | 3 - 4 | 952 | 70 | |
| GenBank mRNA | AY656760.1 | - | 3 - 4 | 891 | 70 | |
More Information
Set Membership: |
|

YY1
Genomic Map