Assay Details
| Gene Symbol: HDHD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968063.1 | XP_027823864.1 | 3 - 4 | 433 | 61 | |
| XM_027968064.1 | XP_027823865.1 | 3 - 4 | 526 | 61 | ||
More Information
Set Membership: |
|

HDHD5
Genomic Map