Assay Details
| Gene Symbol: LOC101117325 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968051.1 | XP_027823852.1 | 16 - 17 | 4017 | 86 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101117325| Gene Symbol: LOC101117325 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968051.1 | XP_027823852.1 | 16 - 17 | 4017 | 86 | |