Assay Details
| Gene Symbol: BCL7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956472.1 | XP_027812273.1 | 2 - 3 | 227 | 88 | |
| XM_027956473.1 | XP_027812274.1 | 2 - 3 | 227 | 88 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BCL7A| Gene Symbol: BCL7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956472.1 | XP_027812273.1 | 2 - 3 | 227 | 88 | |
| XM_027956473.1 | XP_027812274.1 | 2 - 3 | 227 | 88 | ||