Assay Details
| Gene Symbol: BCL7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956472.1 | XP_027812273.1 | 3 - 4 | 333 | 66 | |
| XM_027956473.1 | XP_027812274.1 | 3 - 4 | 333 | 66 | ||
More Information
Set Membership: |
|

BCL7A
Genomic Map