Assay Details
| Gene Symbol: LOC101102529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961780.1 | XP_027817581.1 | 26 - 27 | 2758 | 68 | |
| XM_027961781.1 | XP_027817582.1 | 26 - 27 | 2758 | 68 | ||
| XM_027961783.1 | XP_027817584.1 | 20 - 21 | 2195 | 68 | ||
More Information
Set Membership: |
|

LOC101102529
Genomic Map