Assay Details
| Gene Symbol: FADS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019593.4 | XP_004019642.1 | 2 - 3 | 434 | 63 | |
| XM_012102002.3 | XP_011957392.1 | 2 - 3 | 240 | 63 | ||
| XM_027959823.1 | XP_027815624.1 | 2 - 3 | 278 | 63 | ||
More Information
Set Membership: |
|

FADS1
Genomic Map