Assay Details
| Gene Symbol: LOC101104028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015092453.2 | XP_014947939.1 | 5 - 6 | 1259 | 70 | |
| XM_015092455.2 | XP_014947941.1 | 5 - 6 | 1258 | 70 | ||
More Information
Set Membership: |
|

LOC101104028
Genomic Map