Assay Details
| Gene Symbol: LOC101104028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015092453.2 | XP_014947939.1 | 6 - 7 | 1409 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101104028| Gene Symbol: LOC101104028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015092453.2 | XP_014947939.1 | 6 - 7 | 1409 | 101 | |
Set Membership: |
|