Assay Details
| Gene Symbol: PARP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012095675.3 | XP_011951065.2 | 8 - 9 | 1968 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PARP9| Gene Symbol: PARP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012095675.3 | XP_011951065.2 | 8 - 9 | 1968 | 74 | |
Set Membership: |
|