Assay Details
| Gene Symbol: B4GALT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002689.3 | XP_004002738.1 | 5 - 6 | 936 | 65 | |
| XM_004002690.3 | XP_004002739.1 | 5 - 6 | 952 | 65 | ||
| XM_012184766.3 | XP_012040156.1 | 5 - 6 | 1205 | 65 | ||
More Information
Set Membership: |
|

B4GALT3
Genomic Map