Assay Details
| Gene Symbol: LOC101117577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965980.1 | XP_027821781.1 | 10 - 11 | 1292 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101117577| Gene Symbol: LOC101117577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965980.1 | XP_027821781.1 | 10 - 11 | 1292 | 102 | |
Set Membership: |
|