Assay Details
| Gene Symbol: LOC114113810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965978.1 | XP_027821779.1 | 6 - 7 | 991 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC114113810| Gene Symbol: LOC114113810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965978.1 | XP_027821779.1 | 6 - 7 | 991 | 69 | |
Set Membership: |
|