Assay Details
| Gene Symbol: LOC114116423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973285.1 | XP_027829086.1 | 26 - 27 | 3815 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC114116423| Gene Symbol: LOC114116423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973285.1 | XP_027829086.1 | 26 - 27 | 3815 | 90 | |
Set Membership: |
|