Assay Details
Gene Symbol: MTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976977.1 | XP_027832778.1 | 2 - 3 | 822 | 56 | |
XM_027976978.1 | XP_027832779.1 | 2 - 3 | 252 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
