Assay Details
| Gene Symbol: LOC101104855 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016542.3 | XP_004016591.2 | 5 - 6 | 452 | 73 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101104855| Gene Symbol: LOC101104855 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016542.3 | XP_004016591.2 | 5 - 6 | 452 | 73 | |