Assay Details
| Gene Symbol: FBLN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012098937.3 | XP_011954327.1 | 10 - 11 | 1597 | 61 | |
| XM_012098938.2 | XP_011954328.1 | 11 - 12 | 1491 | 61 | ||
| XM_012098939.3 | XP_011954329.1 | 11 - 12 | 1308 | 61 | ||
| XM_027957332.1 | XP_027813133.1 | 10 - 11 | 1654 | 61 | ||
More Information
Set Membership: |
|

FBLN5
Genomic Map