Assay Details
| Gene Symbol: WASHC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006690.4 | XP_004006739.2 | 28 - 29 | 2975 | 70 | |
| XM_027967682.1 | XP_027823483.1 | 21 - 22 | 2404 | 70 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WASHC4| Gene Symbol: WASHC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006690.4 | XP_004006739.2 | 28 - 29 | 2975 | 70 | |
| XM_027967682.1 | XP_027823483.1 | 21 - 22 | 2404 | 70 | ||