Assay Details
| Gene Symbol: WASHC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006690.4 | XP_004006739.2 | 30 - 31 | 3228 | 67 | |
| XM_027967682.1 | XP_027823483.1 | 23 - 24 | 2657 | 67 | ||
More Information
Set Membership: |
|

WASHC4
Genomic Map