Assay Details
| Gene Symbol: WASHC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006690.4 | XP_004006739.2 | 9 - 10 | 730 | 100 | |
| XM_027967682.1 | XP_027823483.1 | 2 - 3 | 159 | 100 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WASHC4| Gene Symbol: WASHC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006690.4 | XP_004006739.2 | 9 - 10 | 730 | 100 | |
| XM_027967682.1 | XP_027823483.1 | 2 - 3 | 159 | 100 | ||