Assay Details
| Gene Symbol: CERS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969376.1 | XP_027825177.1 | 5 - 6 | 957 | 67 | |
| XM_027969378.1 | XP_027825179.1 | 5 - 6 | 957 | 67 | ||
More Information
Set Membership: |
|

CERS1
Genomic Map