Assay Details
| Gene Symbol: SYBU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973091.1 | XP_027828892.1 | 8 - 9 | 2565 | 58 | |
| XM_027973092.1 | XP_027828893.1 | 7 - 8 | 1056 | 58 | ||
| XM_027973093.1 | XP_027828894.1 | 8 - 9 | 2562 | 58 | ||
| XM_027973094.1 | XP_027828895.1 | 7 - 8 | 2504 | 58 | ||
| XM_027973095.1 | XP_027828896.1 | 7 - 8 | 2501 | 58 | ||
| XM_027973096.1 | XP_027828897.1 | 4 - 5 | 3575 | 58 | ||
More Information
Set Membership: |
|

SYBU
Genomic Map