Assay Details
| Gene Symbol: C15H11orf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979169.1 | XP_027834970.1 | 5 - 6 | 679 | 91 | |
| XM_027979170.1 | XP_027834971.1 | 6 - 7 | 733 | 91 | ||
More Information
Set Membership: |
|

C15H11orf65
Genomic Map