Assay Details
| Gene Symbol: LOC105613829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974353.1 | XP_027830154.1 | 5 - 6 | 1824 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105613829| Gene Symbol: LOC105613829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974353.1 | XP_027830154.1 | 5 - 6 | 1824 | 85 | |
Set Membership: |
|