Assay Details
| Gene Symbol: LOC105607811 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974985.1 | XP_027830786.1 | 2 - 3 | 676 | 91 | |
| XM_027974986.1 | XP_027830787.1 | 2 - 3 | 676 | 91 | ||
| XM_027974988.1 | XP_027830789.1 | 2 - 3 | 675 | 91 | ||
| XM_027974989.1 | XP_027830790.1 | 2 - 3 | 673 | 91 | ||
| XM_027974990.1 | XP_027830791.1 | 3 - 4 | 374 | 91 | ||
More Information
Set Membership: |
|

LOC105607811
Genomic Map