Assay Details
| Gene Symbol: STBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004009927.4 | XP_004009976.1 | 1 - 2 | 375 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

STBD1| Gene Symbol: STBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004009927.4 | XP_004009976.1 | 1 - 2 | 375 | 82 | |
Set Membership: |
|