Assay Details
| Gene Symbol: CNPY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959151.1 | XP_027814952.1 | 3 - 4 | 504 | 61 | |
| XM_027959152.1 | XP_027814953.1 | 3 - 4 | 565 | 61 | ||
More Information
Set Membership: |
|

CNPY3
Genomic Map