Assay Details
| Gene Symbol: AVPI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020087.4 | XP_004020136.2 | 2 - 3 | 608 | 104 | |
| XM_004020088.4 | XP_004020137.2 | 2 - 3 | 967 | 104 | ||
More Information
Set Membership: |
|

AVPI1
Genomic Map