Assay Details
| Gene Symbol: PAGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012104379.3 | XP_011959769.1 | 2 - 3 | 684 | 80 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAGR1| Gene Symbol: PAGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012104379.3 | XP_011959769.1 | 2 - 3 | 684 | 80 | |