Assay Details
Gene Symbol: LOC101108354 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012097075.3 | XP_011952465.1 | 4 - 5 | 719 | 134 | |
XM_012097111.3 | XP_011952501.1 | 4 - 5 | 719 | 134 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
