Assay Details
| Gene Symbol: LOC101108354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012097075.3 | XP_011952465.1 | 5 - 6 | 880 | 68 | |
| XM_012097111.3 | XP_011952501.1 | 5 - 6 | 880 | 68 | ||
More Information
Set Membership: |
|

LOC101108354
Genomic Map