Assay Details
| Gene Symbol: SEPTIN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015094375.2 | XP_014949861.2 | 2 - 3 | 142 | 72 | |
| XM_015094376.2 | XP_014949862.2 | 2 - 3 | 153 | 72 | ||
More Information
Set Membership: |
|

SEPTIN10
Genomic Map