Assay Details
| Gene Symbol: GAL3ST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956773.1 | XP_027812574.1 | 2 - 3 | 190 | 114 | |
| XM_027956774.1 | XP_027812575.1 | 2 - 3 | 375 | 114 | ||
More Information
Set Membership: |
|

GAL3ST1
Genomic Map