Assay Details
Gene Symbol: IFI6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965431.1 | XP_027821232.1 | 3 - 4 | 375 | 90 | |
XM_027965432.1 | XP_027821233.1 | 4 - 5 | 439 | 90 | ||
XM_027965433.1 | XP_027821234.1 | 5 - 6 | 527 | 90 |
